2-Amino-5-chloro-4-methoxybenzoic acid 97%

Supplier: Ambeed
Warning

A241900-100MG A241900-1G
77013-782EA 8 USD
77013-782 77013-784
2-Amino-5-chloro-4-methoxybenzoic acid 97%
2-Amino-5-chloro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MDL Number: MFCD05665825
CAS Number: 79025-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR