2-Octynoic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-octynoate, Methyl oct-2-ynoate

A241870-10G A241870-25G A241870-100G
77000-454EA 20.16 USD
77000-454 77600-954 77672-166
2-Octynoic acid methyl ester 98%
2-Octynoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₁₄O₂
MW: 154.21 g/mol
MDL Number: MFCD00009530
CAS Number: 111-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR