2-Amino-2-(pyridin-2-yl)ethanol dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-2-(2-pyridyl)ethanol Dihydrochloride

A241714-100MG A241714-1G A241714-250MG
77000-450EA 20 USD
77000-450 77000-452 77881-633
2-Amino-2-(pyridin-2-yl)ethanol dihydrochloride 98%
2-Amino-2-(pyridin-2-yl)ethanol dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂Cl₂N₂O
MW: 211.09 g/mol
MDL Number: MFCD12913795
CAS Number: 1187930-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR