Methyl-2-bromo-4-methoxybenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-4-methoxybenzoic acid methyl ester, Methyl 2-bromo-4-methoxybenzoate
A241656-10G
A241656-1G
A241656-250MG
A241656-25G
A241656-5G
A241656-100G
77031-284EA
6.37
USD
77031-284
77031-286
77031-288
77031-290
77031-292
77031-294
77672-164
Methyl-2-bromo-4-methoxybenzoate 98%
Methyl-2-bromo-4-methoxybenzoate
Precaución: for Research Use Only
Formula:
C₉H₉BrO₃ MW: 245.07 g/mol |
MDL Number:
MFCD12026001 CAS Number: 17100-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...