4-Bromo-2-fluoro-3-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A241624-100MG A241624-1G A241624-250MG A241624-5G
77013-774EA 14 USD
77013-774 77013-776 77013-778 77013-780
4-Bromo-2-fluoro-3-methoxybenzoic acid 98%
4-Bromo-2-fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₃
MDL Number: MFCD06656802
CAS Number: 194804-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR