1-Ethynyl-2-methoxy-4-nitrobenzene 97%
Supplier: Ambeed
|
Warning
|
A241421-1G
A241421-250MG
A241421-5G
A241421-100MG
77000-442EA
118
USD
77000-442
77000-444
77000-446
77881-632
1-Ethynyl-2-methoxy-4-nitrobenzene 97%
1-Ethynyl-2-methoxy-4-nitrobenzene
Precaución: for Research Use Only
Formula:
C₉H₇NO₃ |
MDL Number:
MFCD11840985 CAS Number: 1135283-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...