10-Bromo-7H-benzo[c]carbazole 97%

Supplier: Ambeed
Warning

A241249-100MG A241249-1G A241249-250MG A241249-25G A241249-5G
76987-624EA 5.31 USD
76987-624 76987-626 76987-628 76987-630 76987-632
10-Bromo-7H-benzo[c]carbazole 97%
10-Bromo-7H-benzo[c]carbazole

Precaución: for Research Use Only

Formula: C₁₆H₁₀BrN
MW: 296.17 g/mol
MDL Number: MFCD22054961
CAS Number: 1698-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR