5-Bromo-2-pyridineacetic acid 95% Titration by NaOH

Supplier: Ambeed
Warning

Synonyms: 2-(5-Bromopyridin-2-yl)acetic acid

A241033-10G A241033-1G A241033-250MG A241033-5G A241033-25G
77031-276EA 200.52 USD
77031-276 77031-278 77031-280 77031-282 77672-154
5-Bromo-2-pyridineacetic acid 95% Titration by NaOH
5-Bromo-2-pyridineacetic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.034 g/mol
MDL Number: MFCD09999983
CAS Number: 192642-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR