5-Methyl-2,2'-bipyridine 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A240994-100MG A240994-1G A240994-250MG A240994-5G A240994-25G
77013-762EA 19.1 USD
77013-762 77013-764 77013-766 77013-768 77683-730
5-Methyl-2,2'-bipyridine 98%, Ambeed.Inc
5-Methyl-2,2'-bipyridine

Precaución: For research use only.

Formula: C₁₁H₁₀N₂
MW: 170.211 g/mol
MDL Number: MFCD08062569
CAS Number: 56100-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR