N,N-Dimethyl-n-octadecylamine 98%

Supplier: Ambeed
Danger

Synonyms: N,N-Dimethyloctadecan-1-amine, Dymanthine, N,N-Dimethylstearylamine

A240581-25G A240581-100G
77000-410EA 7 USD
77000-410 77672-146
N,N-Dimethyl-n-octadecylamine 98%
N,N-Dimethyl-n-octadecylamine

Precaución: for Research Use Only

Formula: C₂₀H₄₃N
MDL Number: MFCD00048496
CAS Number: 124-28-7
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR