2-(Trifluoromethyl)-D-phenyalanine 98%

Supplier: Ambeed
Warning

A240514-1G A240514-100MG A240514-250MG A240514-5G
77000-408EA 59 USD
77000-408 77600-932 77600-934 77600-936
2-(Trifluoromethyl)-D-phenyalanine 98%
2-(Trifluoromethyl)-D-phenyalanine

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO₂
MDL Number: MFCD01860877
CAS Number: 130930-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR