cis-3,4-Dihydroxypyrrolidine hydrochloride 98%

Supplier: Ambeed
Warning

A240395-1G A240395-250MG A240395-5G
77013-752EA 48 USD
77013-752 77013-754 77013-756
cis-3,4-Dihydroxypyrrolidine hydrochloride 98%
cis-3,4-Dihydroxypyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO₂
MDL Number: MFCD21602660
CAS Number: 186393-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR