(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide 95%

Supplier: Ambeed
Warning

Synonyms: N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide, 4-Formylaminoantipyrine

A240335-250MG A240335-5G
77000-404EA 30.77 USD
77000-404 77000-406
(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide 95%
(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide

Precaución: for Research Use Only

Formula: C₁₂H₁₃N₃O₂
MW: 231.26 g/mol
MDL Number: MFCD00085457
CAS Number: 1672-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR