3-Bromo-2,6-dimethylpyridin-4-amine 98%

Supplier: Ambeed
Warning

A240270-100MG A240270-1G A240270-250MG
77013-746EA 60.48 USD
77013-746 77013-748 77013-750
3-Bromo-2,6-dimethylpyridin-4-amine 98%
3-Bromo-2,6-dimethylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MW: 201.06 g/mol
MDL Number: MFCD24444976
CAS Number: 33259-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR