(2-Amino-4-(trifluoromethyl)thiazol-5-yl)methanol 98%

Supplier: Ambeed
Warning

A240098-100MG A240098-1G A240098-250MG
77013-740EA 28.65 USD
77013-740 77013-742 77013-744
(2-Amino-4-(trifluoromethyl)thiazol-5-yl)methanol 98%
(2-Amino-4-(trifluoromethyl)thiazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₅F₃N₂OS
MW: 198.17 g/mol
MDL Number: MFCD09250889
CAS Number: 1314977-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR