5-(Benzyloxy)-2-bromopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Benzyloxy-2-bromopyridine

A239923-1G A239923-250MG
77013-736EA 94.43 USD
77013-736 77013-738
5-(Benzyloxy)-2-bromopyridine 98%
5-(Benzyloxy)-2-bromopyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MW: 264.11 g/mol
MDL Number: MFCD11874623
CAS Number: 630120-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR