7-Chloro-2-iodothieno[3,2-b]pyridine 95%
Supplier: Ambeed
|
Warning
|
A239839-100MG
A239839-250MG
A239839-5G
A239839-1G
77013-730EA
19
USD
77013-730
77013-732
77013-734
77890-386
7-Chloro-2-iodothieno[3,2-b]pyridine 95%
7-Chloro-2-iodothieno[3,2-b]pyridine
Precaución: for Research Use Only
Formula:
C₇H₃ClINS |
MDL Number:
MFCD08448166 CAS Number: 602303-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...