2-Hydroxy-5-(pyridin-4-yl)benzaldehyde 98%

Supplier: Ambeed
Warning

A239721-250MG A239721-1G
77013-722EA 64.72 USD
77013-722 77683-725
2-Hydroxy-5-(pyridin-4-yl)benzaldehyde 98%
2-Hydroxy-5-(pyridin-4-yl)benzaldehyde

Precaución: For research use only.

Formula: C₁₂H₉NO₂
MW: 199.20 g/mol
MDL Number: MFCD11877749
CAS Number: 342037-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR