Methyl-3-bromo-5-chloro-2-hydroxybenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromo-5-chloro-2-hydroxybenzoate

A239704-10G A239704-1G A239704-250MG A239704-25G A239704-5G
76987-596EA 251.44 USD
76987-596 76987-598 76987-600 76987-602 76987-604
Methyl-3-bromo-5-chloro-2-hydroxybenzoate 97%
Methyl-3-bromo-5-chloro-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₃
MW: 265.5 g/mol
MDL Number: MFCD06203697
CAS Number: 4068-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR