4-Amino-3-chloropyridine 97%

Supplier: Ambeed
Warning

A239661-100G A239661-10G A239661-25G A239661-5G
77000-384EA 206.88 USD
77000-384 77000-386 77000-388 77000-390
4-Amino-3-chloropyridine 97%
4-Amino-3-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD03984473
CAS Number: 19798-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR