4,4'-Bis(2-benzoxazolyl)stilbene 97%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(4-(benzo[d]oxazol-2-yl)phenyl)ethene

A239632-100G A239632-10G A239632-25G A239632-500G A239632-1000G
77000-376EA 21.22 USD
77000-376 77000-378 77000-380 77000-382 77600-930
4,4'-Bis(2-benzoxazolyl)stilbene 97%
4,4'-Bis(2-benzoxazolyl)stilbene

Precaución: for Research Use Only

Formula: C₂₈H₁₈N₂O₂
MW: 414.46 g/mol
MDL Number: MFCD00022846
CAS Number: 1533-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR