2-(5-Fluoro-1H-indol-3-yl)ethanol 98%

Supplier: Ambeed
Warning

A239615-250MG A239615-25G A239615-5G
77013-718EA 13 USD
77013-718 77568-328 77890-383
2-(5-Fluoro-1H-indol-3-yl)ethanol 98%
2-(5-Fluoro-1H-indol-3-yl)ethanol

Precaución: For research use only.

Formula: C₁₀H₁₀FNO
MDL Number: MFCD09752697
CAS Number: 101349-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR