Senicapoc 98%

Supplier: Ambeed
Warning

Synonyms: ICA-17043, ICA17043, 2,2-bis(4-fluorophenyl)-2-phenylacetamide

A239412-100MG A239412-10G A239412-10MG A239412-1MG A239412-50MG A239412-5MG
77031-250EA 106.09 USD
77031-250 77031-252 77031-254 77031-256 77031-258 77031-260
Senicapoc 98%
Senicapoc

Precaución: for Research Use Only

Formula: C₂₀H₁₅F₂NO
MW: 323.34 g/mol
MDL Number: MFCD09027349
CAS Number: 289656-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR