6-Bromo-2,3-dihydro-1H-indene-1-carboxylic acid 97%

Supplier: Ambeed
Warning

A239261-100MG A239261-10G A239261-1G A239261-250MG
77013-708EA 48.81 USD
77013-708 77013-710 77013-712 77013-714
6-Bromo-2,3-dihydro-1H-indene-1-carboxylic acid 97%
6-Bromo-2,3-dihydro-1H-indene-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MW: 241.08 g/mol
MDL Number: MFCD10697921
CAS Number: 52651-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR