4-Bromo-1-methyl-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A238906-100MG A238906-1G A238906-250MG A238906-5G
76987-576EA 14.86 USD
76987-576 76987-578 76987-580 76987-582
4-Bromo-1-methyl-1H-pyrrolo[2,3-b]pyridine 95%
4-Bromo-1-methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.06 g/mol
MDL Number: MFCD12964073
CAS Number: 1234616-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR