Fmoc-4-nitro-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-p-nitro-D-Phe-OH, Fmoc-D-Phe(4-NO₂)-OH, Fmoc-D-Phe(4-NO2)-OH

A238512-100G A238512-25G A238512-5G A238512-1G A238512-250MG
77000-356EA 197 USD
77000-356 77000-358 77000-360 77672-136 77881-625
Fmoc-4-nitro-D-phenylalanine 97%
Fmoc-4-nitro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀N₂O₆
MW: 432.432 g/mol
MDL Number: MFCD00237029
CAS Number: 177966-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR