(2R,5R)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 5-hydroxy-1,3-oxathiolane-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (1R,2S,5R)-Menthol-5R-hydroxy-[1,3]-oxathiolane-2R-carboxylate

A238462-10G A238462-5G A238462-25G
77000-352EA 60.48 USD
77000-352 77000-354 77672-132
(2R,5R)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 5-hydroxy-1,3-oxathiolane-2-carboxylate 97%
(2R,5R)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 5-hydroxy-1,3-oxathiolane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₄O₄S
MW: 288.4 g/mol
MDL Number: MFCD08273790
CAS Number: 147126-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR