7-Bromochroman 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A238227-100MG A238227-250MG A238227-5G A238227-1G
77013-688EA 48.81 USD
77013-688 77013-690 77013-692 77683-712
7-Bromochroman 97%, Ambeed.Inc
7-Bromochroman

Precaución: For research use only.

Formula: C₉H₉BrO
MW: 213.07 g/mol
MDL Number: MFCD22536189
CAS Number: 941710-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR