(3-Methoxypyridin-2-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A238185-1G A238185-250MG
77013-682EA 789 USD
77013-682 77013-684 77013-686
(3-Methoxypyridin-2-yl)methanamine hydrochloride 95%
(3-Methoxypyridin-2-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂O
MDL Number: MFCD27923105
CAS Number: 1588441-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR