3-Cyclopropyl-3-oxopropanenitrile 95%

Supplier: Ambeed
Danger

A237863-10G A237863-5G A237863-25G
77013-678EA 43 USD
77013-678 77013-680 77890-378
3-Cyclopropyl-3-oxopropanenitrile 95%
3-Cyclopropyl-3-oxopropanenitrile

Precaución: for Research Use Only

Formula: C₆H₇NO
MDL Number: MFCD04114392
CAS Number: 118431-88-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR