2-(4-(2-(Trifluoromethyl)benzoyl)piperazin-1-yl)thiazole-5-carboxamide 97%

Supplier: Ambeed
Warning

A237812-100MG A237812-1G A237812-250MG
76987-540EA 26 USD
76987-540 76987-542 76987-544
2-(4-(2-(Trifluoromethyl)benzoyl)piperazin-1-yl)thiazole-5-carboxamide 97%
2-(4-(2-(Trifluoromethyl)benzoyl)piperazin-1-yl)thiazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₁₆H₁₅F₃N₄O₂S
MW: 384.38 g/mol
MDL Number: MFCD13193183
CAS Number: 916888-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR