4-(4-Methyl-1,3-dioxolan-2-yl)-1,2-methylenedioxybenzene 98% (by GC)

Supplier: Ambeed
Warning

A237659-10G A237659-25G
77013-674EA 13 USD
77013-674 77013-676
4-(4-Methyl-1,3-dioxolan-2-yl)-1,2-methylenedioxybenzene 98% (by GC)
4-(4-Methyl-1,3-dioxolan-2-yl)-1,2-methylenedioxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MDL Number: MFCD13194964
CAS Number: 61683-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR