Fmoc-Thr(Bzl)-OH 95%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-O-benzyl-L-threonine

A237589-100G A237589-1G A237589-5G A237589-25G A237589-10G
77000-336EA 132 USD
77000-336 77000-338 77000-340 77672-124 77881-620
Fmoc-Thr(Bzl)-OH 95%
Fmoc-Thr(Bzl)-OH

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₅
MDL Number: MFCD00077073
CAS Number: 117872-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR