4-(2-Pyridyl)-3-butyn-1-ol 95%

Supplier: Ambeed
Warning

A237506-10G A237506-1G A237506-250MG A237506-5G
77013-668EA 662 USD
77013-668 77013-670 77013-672 77890-373
4-(2-Pyridyl)-3-butyn-1-ol 95%
4-(2-Pyridyl)-3-butyn-1-ol

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD18384524
CAS Number: 395652-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR