Methyl (2R,3S)-3-(tert-butoxycarbonylamino)-2-hydroxy-3-phenylpropionate 97%

Supplier: Ambeed
Warning

A237411-250MG A237411-5G
77000-332EA 8.49 USD
77000-332 77000-334
Methyl (2R,3S)-3-(tert-butoxycarbonylamino)-2-hydroxy-3-phenylpropionate 97%
Methyl (2R,3S)-3-(tert-butoxycarbonylamino)-2-hydroxy-3-phenylpropionate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₅
MW: 295.33 g/mol
MDL Number: MFCD09037385
CAS Number: 124605-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR