1-(4-Hydroxyphenyl)-1H-tetrazole 95%

Supplier: Ambeed
Danger

Synonyms: 4-(1H-Tetrazol-1-yl)phenol

A237175-10G A237175-5G
77031-206EA 956.94 USD
77031-206 77031-208
1-(4-Hydroxyphenyl)-1H-tetrazole 95%
1-(4-Hydroxyphenyl)-1H-tetrazole

Precaución: for Research Use Only

Formula: C₇H₆N₄O
MW: 162.1500 g/mol
MDL Number: MFCD03490124
CAS Number: 64001-11-2
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR