N-Benzoyl-N-phenylhydroxylamine 98%

Supplier: Ambeed
Warning

Synonyms: Tantalon, N-Phenylbenzohydroxamic Acid, N-Hydroxy-N-phenylbenzamide

A236968-1G A236968-250MG
77000-320EA 9.55 USD
77000-320 77000-322
N-Benzoyl-N-phenylhydroxylamine 98%
N-Benzoyl-N-phenylhydroxylamine

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
MDL Number: MFCD00002111
CAS Number: 304-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR