Phenethyl methanesulfonate 98%

Supplier: Ambeed
Warning

A236835-1G A236835-5G
76987-528EA 123.07 USD
76987-528 77683-704
Phenethyl methanesulfonate 98%
Phenethyl methanesulfonate

Precaución: for Research Use Only

Formula: C₉H₁₂O₃S
MW: 200.26 g/mol
MDL Number: MFCD01715258
CAS Number: 20020-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR