4-Amino-6-iodoindole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Iodo-1H-indol-4-amine

A236720-250MG A236720-1G
76987-522EA 155.96 USD
76987-522 77683-703
4-Amino-6-iodoindole 98%
4-Amino-6-iodoindole

Precaución: For research use only.

Formula: C₈H₇IN₂
MW: 258.059 g/mol
MDL Number: MFCD07781458
CAS Number: 885520-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR