4-Chloro-2,5-dimethoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A236680-1G A236680-250MG
77013-664EA 353.28 USD
77013-664 77013-666
4-Chloro-2,5-dimethoxybenzaldehyde 95%
4-Chloro-2,5-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
MDL Number: MFCD12964185
CAS Number: 90064-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR