2-(Hydroxymethyl)-2-nitro-1,3-propanediol 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Hydroxymethyl)-2-nitropropane-1,3-diol, Tris(hydroxymethyl)nitromethane, Trimethylolnitromethane
A236261-1000G
A236261-100G
A236261-25G
A236261-500G
77000-312EA
109.28
USD
77000-312
77000-314
77000-316
77000-318
2-(Hydroxymethyl)-2-nitro-1,3-propanediol 99+%
2-(Hydroxymethyl)-2-nitro-1,3-propanediol
Precaución: for Research Use Only
Formula:
C₄H₉NO₅ MW: 151.12 g/mol |
MDL Number:
MFCD00007395 CAS Number: 126-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...