1-(2-Chlorophenyl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A236099-100MG A236099-1G
76987-516EA 22 USD
76987-516 76987-518
1-(2-Chlorophenyl)ethanamine hydrochloride 95%
1-(2-Chlorophenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClN·ClH
MDL Number: MFCD11841333
CAS Number: 856629-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR