4-(9H-Carbazol-9-yl)benzaldehyde 95%

Supplier: Ambeed
Warning

A236022-1G A236022-250MG A236022-5G
76987-510EA 19 USD
76987-510 76987-512 76987-514
4-(9H-Carbazol-9-yl)benzaldehyde 95%
4-(9H-Carbazol-9-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₉H₁₃NO
MDL Number: MFCD19441374
CAS Number: 110677-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR