3-Chloropyrazin-2(1H)-one 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A235959-100MG A235959-250MG A235959-1G A235959-5G
76987-506EA 43.5 USD
76987-506 76987-508 77593-516 77683-697
3-Chloropyrazin-2(1H)-one 95%, Ambeed.Inc
3-Chloropyrazin-2(1H)-one

Precaución: For research use only.

Formula: C₄H₃ClN₂O
MW: 130.53 g/mol
MDL Number: MFCD18427098
CAS Number: 105985-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR