Alizarin complexone 95%

Supplier: Ambeed
Warning

Synonyms: Alizarin-3-methyliminodiacetic acid, 2,2'-(((3,4-Dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl)azanediyl)diacetic acid, 3,4-dihydroxyanthraquinon-2-ylmethyliminodi(acetic acid), Alizarine-3-methylamine-N,N-diacetic acid

A235856-1G A235856-250MG A235856-25G A235856-5G
77031-190EA 13.8 USD
77031-190 77031-192 77031-194 77031-196
Alizarin complexone 95%
Alizarin complexone

Precaución: for Research Use Only

Formula: C₁₉H₁₅NO₈
MW: 385.33 g/mol
MDL Number: MFCD00001202
CAS Number: 3952-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR