3-Aminopiperidine hydrochloride, N1-CBZ protected 97%

Supplier: Ambeed
Warning

A235752-10G A235752-1G A235752-250MG
77013-646EA 255 USD
77013-646 77013-648 77013-650
3-Aminopiperidine hydrochloride, N1-CBZ protected 97%
3-Aminopiperidine hydrochloride, N1-CBZ protected

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂·ClH
MDL Number: MFCD04038445
CAS Number: 960541-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR