3-Amino-6-chloro-2-pyrazinecarbonitrile 95%

Supplier: Ambeed
Warning

A235366-1G A235366-250MG
77031-178EA 79.57 USD
77031-178 77031-180
3-Amino-6-chloro-2-pyrazinecarbonitrile 95%
3-Amino-6-chloro-2-pyrazinecarbonitrile

Precaución: for Research Use Only

Formula: C5H3ClN4
MW: 154.5600 g/mol
MDL Number: MFCD10697784
CAS Number: 17231-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR