Boc-L-β-homoglutamic acid-d-benzyl ester 98%

Supplier: Ambeed
Warning

Synonyms: (S)-6-(Benzyloxy)-3-((tert-butoxycarbonyl)amino)-6-oxohexanoic acid, Boc-β-HomoGlu(OBzl)-OH, Boc-β-HoGlu(OBzl)-OH

A235303-100MG A235303-1G A235303-250MG A235303-5G
77000-292EA 20 USD
77000-292 77000-294 77000-296 77672-096
Boc-L-β-homoglutamic acid-d-benzyl ester 98%
Boc-L-beta-homoglutamic acid-d-benzyl ester

Precaución: for Research Use Only

Formula: C₁₈H₂₅NO₆
MW: 351.4 g/mol
MDL Number: MFCD01862936
CAS Number: 218943-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR