Phenyl-1,4-dihydroxy-2-naphthoate 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Dihydroxy-2-naphthoic Acid Phenyl Ester, Phenyl 1,4-dihydroxy-2-naphthoate

A235257-25G A235257-5G
77031-168EA 115.64 USD
77031-168 77031-170
Phenyl-1,4-dihydroxy-2-naphthoate 98%
Phenyl-1,4-dihydroxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₇H₁₂O₄
MW: 280.28 g/mol
MDL Number: MFCD00075770
CAS Number: 54978-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR