3-Acetylquinoline 97%

Supplier: Ambeed
Warning

A235154-100MG A235154-1G A235154-250MG A235154-5G A235154-25G
77000-284EA 21 USD
77000-284 77000-286 77000-288 77000-290 77881-613
3-Acetylquinoline 97%
3-Acetylquinoline

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MW: 171.20 g/mol
MDL Number: MFCD00030698
CAS Number: 33021-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR